Learn More
Tyrosinase Sheep anti-Fungi, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen PA185947
Catalog No. PA185947
Add to cart
Description
Description
Sheep anti Mushroom tyrosinase antibody recognizes mushroom tyrosinase, also known as 5,6-dihydroxyindole-2-carboxylic acid oxidase, DHICA oxidase, Glycoprotein 75, Melanoma antigen gp75, Tyrosinase-related protein 1 or TRP-1.
Tyrosinase is a copper-containing metalloglycoprotein that catalyzes several steps in the melanin pigment biosynthetic pathway; the hydroxylation of tyrosine to L-3,4-dihydroxy-phenylalanine (dopa), and the subsequent oxidation of dopa to dopaquinone. Mutations of the tyrosinase gene occur in various forms of albinism. Tyrosinase is one of the targets for cytotoxic T-cell recognition in melanoma patients.
Specifications
Specifications
Tyrosinase
Polyclonal
whole serum with 0.09% sodium azide
OCA1A; OCAIA; SHEP3
Highly pure tyrosinase from mushrooms.
200 μL
4°C or -20°C if preferred
Primary
Fungi
ELISA, Western Blot
Unconjugated
Liquid
Sheep
IgG
RUO
Antibody
Polyclonal