Learn More
MilliporeSigma™ anti-Tyrosine Hydroxylase, Polyclonal, (Chemicon) Non-distribution product as customer accommodation.
Manufacturer:  MilliporeSigma™ AB9702/DEL
Catalog No. 50-174-228
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Tyrosine hydroxylase is involved in the conversion of tyrosine to dopamine. As the rate-limiting enzyme in the synthesis of catecholamines, tyrosine hydroxylase has a key role in the physiology of adrenergic neurons.
Immunocytochemistry, Immunohistochemistry, Western Blotting
Specifications
Specifications
Tyrosine Hydroxylase
Unconjugated
Chicken
IgY
RUO
Primary
Human, Murine
0.2mg/mL
Affinity Purified
Synthetic peptides from human/mouse Tyrosine Hydroxylase.
500μL
Neuroscience
Polyclonal