Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Tyrosine Hydroxylase, Polyclonal, Novus Biologicals™

Specifications

Host Species Rabbit
Immunogen SDS-denatured tyrosine hydroxylase purified from pheochromocytoma of Rat
Antigen Tyrosine Hydroxylase
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227146
View Documents
Novus Biologicals
NBP227146
0.1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Specifications

Specifications

Rabbit
Tyrosine Hydroxylase
Affinity Purified
RUO
The antibody recognizes all mammalian and at least some non-mammalian forms of the enzyme in Western blots and in IHC/IF.
0.1mL in 10mM HEPES (pH 7.5), 150mM NaCl, 0.1mg/mL BSA and 50% glycerol. Adequate amount of material to conduct 10 mini Western Blots.
SDS-denatured tyrosine hydroxylase purified from pheochromocytoma of Rat
Polyclonal
Primary
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
7054
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit