Learn More
TrkC Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA118404
Catalog No. 01-670-654
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-18404 detects Tyrosine Kinase Receptor C from human, mouse, and rat samples. PA1-18404 has been successfully used in immunohistochemistry procedures. The PA1-18404 immunogen is the extracellular domain of glycosylated human TrkC protein produced in CHO cells. Reconstitute with 100 μL of distilled water.
This gene encodes a member of the neurotrophic tyrosine receptor kinase (NTRK) family. This kinase is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. Signalling through this kinase leads to cell differentiation and may play a role in the development of proprioceptive neurons that sense body position. Mutations in this gene have been associated with medulloblastomas, secretory breast carcinomas and other cancers.
Specifications
Specifications
TrkC
Polyclonal
whole serum with no preservative
Q16288, Q6VNS1
TrkC
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry
Unconjugated
NTRK3
Lyophilized
NTRK3
Extracellular domain of glycosylated human TrkC protein produced in CHO cells.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
18213, 4916