Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TYSND1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA523587
Description
Specifications
TYSND1 | |
Polyclonal | |
TYSND1 | |
NET41, peroxisomal leader peptide-processing protease, trypsin domain-containing protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
219743, 71767 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q2T9J0, Q9DBA6 | |
TYSND1 | |
KLH conjugated synthetic peptide between 522-550 amino acids from the C-terminal region of human TYSND1 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction