Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TYW3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521154

Catalog No. PIPA521154


Add to cart

Description

Description

A suggested positive control is A549 cell lysate. PA5-21154 can be used with blocking peptide PEP-1268.

TYW3 is an enzyme that participates in the wybutosine-tRNA (Phe) biosynthesis pathway. Wybutosine (yW) is a hypermodified guanosine at the 3-prime position adjacent to the anticodon of phenylalanine tRNA that stabilizes codon-anticodon interactions during decoding on the ribosome. TYW3 is involved in a multistep enzymatic reaction that stabilizes codon-anticodon base-pairing during the ribosomal decoding process, thereby ensuring correct translation.
Specifications

Specifications

TYW3
Polyclonal
Unconjugated
TYW3
Liquid
TYW3
A 17 amino acid peptide near the center of human TYW3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q6IPR3, Q8BSA9
C1orf171; tRNA wybutosine-synthesizing protein 3 homolog; tRNA-yW-synthesizing protein 3
Rabbit
IgG
100 μg
4° C
Primary
127253, 209584
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit