Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-U2AF35, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP119120
Description
U2AF35 Polyclonal specifically detects U2AF35 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
U2AF35 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
FP793, human U2 snRNP auxiliary factor small subunit10U2AF35DKFZp313J1712, RN, RNU2AF1,35-KD subunit, U2 auxiliary factor 35 kDa subunit, U2 small nuclear RNA auxiliary factor 1, U2 small nuclear RNA auxillary factor 1, U2 snRNP auxiliary factor small subunit | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse. Based on 100% sequence identity, this antibody is predicted to react with Rat, Zebrafish, X. laevis, X. tropicalis, Chicken, Turkey, Bovine, Dog, Horse, Guinea pig_10141, Panda, Orangutan, Rhesus Monkey, Chimpanzee, Water flea, Black-legged tick, Purple sea urchin, Florida lancelet and Soft tick. | |
Store at 4C. Do not freeze. |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunohistochemistry, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:10-1:500 | |
NP_006749.1 | |
U2AF1 | |
The immunogen recognized by this antibody maps to a region between residue 140 and 190 of human U2 small nuclear ribonucleoprotein auxillary factor, 35-KD subunit using the numbering given in entry NP_006749.1 (GeneID 7307). | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
7307 | |
Human, Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only