Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-UAP1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP132823

Catalog No. NBP132823


Add to cart

Description

Description

UAP1 Polyclonal antibody specifically detects UAP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

UAP1
1.0mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
UAP1
Recombinant protein encompassing a sequence within the center region of human UAP1. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Primary
6675
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
AGX, AGX1, AGX2, Antigen X, EC 2.7.7, EC 2.7.7.23, EC 2.7.7.27, SPAG2, sperm associated antigen 2, Sperm-associated antigen 2, UDP-N-acetylhexosamine pyrophosphorylase, UDP-N-acteylglucosamine pyrophosphorylase 1
Rabbit
IgG
100 ul
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only