Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBAC1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525271

Catalog No. PIPA525271


Add to cart

Description

Description

UBAC1 belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein has two RRM domains that bind to RNAs.
Specifications

Specifications

UBAC1
Polyclonal
PBS with 0.09% sodium azide
Q9BSL1
GBDR1, RP11-432J22.3, UBADC1, E3 ubiquitin-protein ligase subunit KPC2, UBA domain-containing protein 1, glialblastoma cell differentiation-related protein 1, kip1 ubiquitination-promoting complex protein 2, putative glialblastoma cell differentiation-related protein, ubiquitin associated
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10422
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
UBAC1
Liquid
UBAC1
KLH conjugated synthetic peptide between 264-292 amino acids from the C-terminal region of human UBAC1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit