Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBAC2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525216

Catalog No. PIPA525216

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with chicken and non-human primate based on sequence homology.

Specifications

Specifications

UBAC2
Polyclonal
Unconjugated
UBAC2
Liquid
UBAC2
KLH conjugated synthetic peptide between 63-91 amino acids from the N-terminal region of human UBAC2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q8NBM4, Q8R1K1
PHGDHL1, RP11-178C10.1, UBA domain-containing protein 2, phosphoglycerate dehydrogenase like 1, phosphoglycerate dehydrogenase-like protein 1, ubiquitin-associated domain-containing protein 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
337867, 68889
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit