Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ubiquitin-conjugating enzyme E2B (RAD6 homolog), Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human UBE2B protein.

Manufacturer:  Abnova Corporation H00007320B01

Catalog No. 89-006-414


Add to cart

Description

Description

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is required for post-replicative DNA damage repair. Its protein sequence is 100% identical to the mouse, rat, and rabbit homologs, which indicates that this enzyme is highly conserved in eukaryotic evolution. [provided by RefSeq

Sequence: MSTPARRRLMRDFKRLQEDPPVGVSGAPSENNIMQWNAVIFGPEGTPFEDGTFKLVIEFSEEYPNKPPTVRFLSKMFHPNVYADGSICLDILQNRWSPTYDVSSILTSIQSLLDEPNPNSPANSQAAQLYQENKREYEKRVSAIVEQSWNDS
Specifications

Specifications

ubiquitin-conjugating enzyme E2B (RAD6 homolog)
Unconjugated
No additive
BC008470
E2-17kDa|HHR6B|HR6B|RAD6B|UBC2
Mouse
50 uL
Ubiquitin/Proteasome
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human UBE2B protein.
UBE2B
Serum
UBE2B
UBE2B (AAH08470, 1 a.a. ~ 152 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
7320
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit