Learn More
UBE2E3 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of UBE2E3.
Manufacturer: Abnova Corporation PAB1771
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein shares 100% sequence identity with the mouse and rat counterparts, which indicates that this enzyme is highly conserved in eukaryotes. Two alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Specifications
UBE2E3 | |
Unconjugated | |
Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
UBE2E3 | |
UBCH9|UbcM2 | |
Rabbit | |
Protein G purification | |
RUO | |
Primary | |
10477 |
Immunohistochemistry (PFA fixed), Western Blot | |
Rabbit polyclonal antibody raised against synthetic peptide of UBE2E3. | |
In PBS (0.09% sodium azide) | |
Liquid | |
UBE2E3 | |
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human UBE2E3. | |
400 uL | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
Human, Murine |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only