Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2J1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178065

Catalog No. NBP178065


Add to cart

Description

Description

UBE2J1 Polyclonal antibody specifically detects UBE2J1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation.
Specifications

Specifications

UBE2J1
0.71 mg/ml
Western Blot 1:200-1:2000, ELISA 1:1000-1:5000, Immunoprecipitation 1-2 ug
Q9Y385
CGI-76, EC 6.3.2.19, HSPC153, HSPC205, HSU93243, HsUBC6e, MGC12555, NCUBE-1, NCUBE1HSUBC6e, Non-canonical ubiquitin-conjugating enzyme 1, non-canonical ubquitin conjugating enzyme 1, Ubc6p, ubiquitin-conjugating enzyme E2 J1, ubiquitin-conjugating enzyme E2, J1 (UBC6 homolog, yeast), Yeast ubiquitin-conjugating enzyme UBC6 homolog E
Rabbit
IgG
0.1 mg
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
ELISA, Immunoprecipitation, Western Blot
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Affinity Purified
UBE2J1
This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to residues near the amino terminus of the human UBE2J1 protein.
Immunogen affinity purified
RUO
Primary
51465
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only