Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2L6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511969

Catalog No. PIPA511969

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s) and ubiquitin-protein ligases (E3s). UBE2L6 is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is demonstrated to function in the E6/E6-ap-induced ubiquitination of p53.
Specifications

Specifications

UBE2L6
Polyclonal
PBS with 0.09% sodium azide
O14933
Ubiquitin/ISG15-conjugating enzyme E2 L6; Ubiquitin-protein ligase L6; Ubiquitin carrier protein L6; UbcH8; Retinoic acid-induced gene B protein; RIG-B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9246
Immunohistochemistry, Western Blot
Unconjugated
UBE2L6
Liquid
UBE2L6
KLH conjugated synthetic peptide between 123-153 amino acids from the C-terminal region of human UBE2L6
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit