Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBC13 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141188

Catalog No. 01-675-753


Add to cart

Description

Description

Suggested positive control: antigen standard for UBE2N (transient overexpression lysate), human heart tissue, human heart protein.

UBE2N (ubiquitin-conjugating enzyme E2N) is a novel human ubiquitin-conjugating enzyme with an ability to form a thiol ester linkage with ubiquitin in an ubiquitin-activating enzyme E1-dependent manner. The amino acid sequence showed 80% identity with the Drosophila ft.s bendless gene product (ubiquitin-conjugating enzyme E2). The corresponding transcripts are highly expressed in heart, skeletal muscle, and testis. UBE2N protein may be involved in protein degradation mainly in the muscles and testis.
Specifications

Specifications

UBC13
Polyclonal
Unconjugated
UBE2N
Liquid
UBE2N
Synthetic peptides corresponding to residues 2-19 and 131-148 of human UBE2N.
Protein G
RUO
Antibody
Polyclonal
Human
Immunoprecipitation, Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P61088
MGC131857, MGC8489, UBC13, UbcH-ben
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
7334
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit