Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2V1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141381

Catalog No. 01-675-510


Add to cart

Description

Description

Suggested positive control: antigen standard for UBE2V1 (transient overexpression lysate), Jurkat, HeLa or RAW cell lysates.

The E2 enzyme, Ubc13, and the E2 enzyme variants, Uevs, form stable, high affinity complexes for the assembly of Lys63-linked ubiquitin chains. This complex has been shown to activate NF-kB signaling pathway by linking TRAF6 to IKKs. Blockade of this polyubiquitin chain synthesis, but not inhibition of the proteasome, prevents the activation of IKK by TRAF6. These results suggest a new regulatory function for ubiquitin, in which IKK is activated through the assembly of Lys63-linked polyubiquitin chains. The UEV proteins are also involved in the control of differentiation.
Specifications

Specifications

UBE2V1
Polyclonal
Unconjugated
UBE2V1
Liquid
UBE2V1
Synthetic peptide corresponding to residues 31-49 of human UBE2V1a.
Protein G
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.2% gelatin and 0.05% sodium azide
Q13404
CROC1, TRAF6-regulated IKK activator 1 beta, Uev1A, CROC-1UBE2V, ubiquitin-conjugating enzyme E2 variant 1, UEV1A, UEV-1CIR1, UEV1DNA-binding protein
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
7335
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit