Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-UBE4A, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232199

Catalog No. NBP232199


Add to cart

Description

Description

UBE4A Polyclonal antibody specifically detects UBE4A in Human samples. It is validated for Immunoprecipitation.
Specifications

Specifications

UBE4A
1.0 mg/mL
Immunoprecipitation 2 - 10 ug/mg lysate
NP_001191006.1
E4MGC133315, KIAA0126ubiquitination factor E4A (homologous to yeast UFD2), ubiquitination factor E4A (UFD2 homolog, yeast), UBOX2, UFD2ubiquitin conjugation factor E4 A
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
UBE4A
The immunogen this antibody was made to, maps to a region between residue 600 to 650 of human Ubiquitination Factor E4A using the numbering given in entry NP_001191006.1 (GeneID 9354).
Immunogen affinity purified
RUO
Primary
9354
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only