Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBIAD1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $409.50
Specifications
Antigen | UBIAD1 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100468SS
|
Novus Biologicals
NB100468SS |
0.025 mL |
Each for $204.00
|
|
|||||
NB100468
|
Novus Biologicals
NB100468 |
0.1 mL |
Each for $409.50
|
|
|||||
Description
UBIAD1 Polyclonal specifically detects UBIAD1 in Human samples. It is validated for Western Blot.Specifications
UBIAD1 | |
Western Blot 1:500 | |
Polyclonal | |
Rabbit | |
Cancer | |
PBS with 0.02% Sodium Azide | |
EC 2.5.1.-, RP4-796F18.1, SCCD, Schnyder crystalline corneal dystrophy, TERE1transitional epithelia response protein, Transitional epithelial response protein 1, UbiA prenyltransferase domain containing 1, ubiA prenyltransferase domain-containing protein 1 | |
UBIAD1 | |
IgG | |
Affinity Purified | |
This is specific for human TERE1. |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
Q9Y5Z9 | |
29914 | |
A synthetic peptide made to residues 32-45 [PEQDRLPQRSWRQK] of the 37 kDa human TERE1 protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
38 kDa |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title