Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ubiquilin 1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA512079
Description
This antibody is predicted to react with rat based on sequence homology.
PLIC1 is an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases, and thus are thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This ubiquilin has also been shown to modulate accumulation of presenilin proteins, and is found in lesions associated with Alzheimer′s and Parkinson′s disease.Specifications
Ubiquilin 1 | |
Polyclonal | |
UBQLN1 | |
Ubiquilin 1; Protein linking IAP with cytoskeleton-1; PLIC-1; hPLIC-1; UBQLN1 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
29979, 56085 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q9UMX0, Q8R317 | |
UBQLN1 | |
KLH conjugated synthetic peptide between 296-326 amino acids from the central region of human Ubiquilin1 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title