Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PGP9.5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA110024
Description
PA1-10024 was made against full length recombinant human UCHL1 expressed in and purified from E. coli and can be used to identify neurons and their processes in culture or in sections. The antibody works cleanly on appropriate lysates of cell and tissues.
PGP9.5 Polyclonal antibody specifically detects PGP9.5 in Bovine, Chicken, Equine, Human, Mouse, Porcine, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
PGP9.5 | |
Polyclonal | |
Unconjugated | |
Uchl1 | |
AW822034; C88048; epididymis luminal protein 117; epididymis secretory protein Li 53; gad; gracile axonal dystrophy; HEL-117; HEL-S-53; MGC133697 protein; NDGOA; neuron cytoplasmic protein 9.5; OTTHUMP00000218137; OTTHUMP00000218139; OTTHUMP00000218140; OTTHUMP00000218141; PARK5; PGP 9.5; PGP9.5; PGP95; protein gene product 9.5; R75593; ubiquitin carboxyl-terminal esterase; ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase); ubiquitin carboxyl-terminal hydrolase isozyme L1; ubiquitin carboxyl-terminal hydrolase L1; ubiquitin carboxy-terminal hydrolase L1; ubiquitin C-terminal hydrolase; ubiquitin C-terminal hydrolase 1; ubiquitin C-terminal hydrolase L1; ubiquitin thioesterase L1; ubiquitin thiolesterase; ubiquitin thiolesterase L1; UCHL; UCHL1; UCH-L1; UCHL-1 | |
Rabbit | |
100 μL | |
Primary | |
Bovine, Chicken, Equine, Human, Mouse, Porcine, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
Conc. Not Determined | |
whole serum with 5mM sodium azide | |
P09936, P23356, Q00981, Q6SEG5, Q9GM50, Q9R0P9 | |
Uchl1 | |
Human UCHL1 expressed in and purified from E. coli. | |
RUO | |
100033838, 22223, 29545, 396637, 514394, 7345, 770302 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction