Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ubiquitin Mouse anti-Vertebrates, Clone: HWA4C4, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051313
Description
Anti-Ubiquitin Antibody, Lys63-Specifimmunocytochemistry, clone HWA4C4 is an antibody against Ubiquitin for use in ELISA, immunohistochemistry, IC and western blotting.Specifications
Ubiquitin | |
Monoclonal | |
Unconjugated | |
NM_018955.2 | |
Mouse | |
100 μg | |
Vertebrates | |
Purified |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
HWA4C4 | |
Purified from ascites by protein G chromatography. Supplied at 1mg/mL in 0.1M Tris-Glycine (pH7.4) 150mM NaCl, 0.05% sodium azide. | |
UBB, UBC, FLJ25987, MGC8385 , OTTHUMP00000064960, OTTHUMP00000064961, UBA80, UBCEP1, UBCEP2 | |
Immunogen used was a branched peptide corresponding to the Lys63 ubiquitination site. The C-terminus of Ubiquitin 71-76 peptide (RLRGG) was attached to the K63 in the Ubiquitin 58-63 peptide (CDYNIQKEST). | |
Primary | |
2-8°C for one year | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only