Learn More
MilliporeSigma™ anti-Ubiquitin, Clone: P4D1-A11, ™ (Upstate™) Non-distribution product as customer accommodation.
Manufacturer:  MilliporeSigma™ 05944/DEL
Catalog No. 50-171-915
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Hochstrasser, M. Annu. Rev. Genet., 30: 405-39, 1996. Laney, J.D., and Hochstrasser, M. Methods Enzymol., 351: 248-257, 2002.Gao, M., and Karin, M. Mol. Cell, 19: 581-593, 2005. Hershko, A. Cell Death Differ., 12: 1191-1197, 2005.
Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
Ubiquitin
Semi-purified
Total bovine ubiquitin
100μg
Protein Trafficking
Monoclonal
Unconjugated
Murine
IgG
RUO
Primary
Human, Yeast