Learn More
Ubiquitin, clone: Ubi-1, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110035
Catalog No. 01-670-292
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Ubiquitin is a highly conserved globular 76 amino acid protein of about 8.5kDa molecular weight. It has an important role in the targeting of proteins for proteolytic degradation. Proteins to be degraded are covalantly coupled to the C-terminus of ubiquitin by means of ubiquitin ligases. The ubiquitin itself is frequently also ubiquitinated, producing a polyubiquitin chain.
This antibody was orginally validated as part of a Thermo Scientific Cellomics High Content Screening Kit. The antibody sold separately may have slightly different performance and may need to be further optimized for the best results.
Specifications
Specifications
Ubiquitin
Monoclonal
Unconjugated
RPS27A
Ascites
RPS27A
Purified ubiquitin conjugated with glutaroldehyde to KLH
100μL
Store at 4°C short term. For long term storage, Store at -20°C, avoiding freeze/thaw cycles.
Primary
6233
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Western Blot
Ubi-1
ascites with no preservative
P62992, P79781, P15357, P62979, P62983, P62982
60S ribosomal protein L40, HUBCEP52, RPL40, Ubiquitin 52 amino acid fusion protein, Ubiquitin A52 residue ribosomal protein fusion product 1, Ubiquiti
Murine
IgG1, κ
RUO
Antibody
Monoclonal
Bovine, C. elegans, Chicken, Drosophila, Equine, Human, Murine, Porcine, Rat