Learn More
MilliporeSigma™ anti- Ubiquitin, Lys48-Specific, Clone: Apu2 Rabbit Monoclonal,
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma™ 051307AF488
Catalog No. 05-130-7AF488
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifically detects Ubiquitin, Lys48-Specific, Clone: Apu2 in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry
Polyubiquitin-B (UniProt P0CG47) is encoded by the UBB gene (Gene ID 7314) in human. Ubiqutin (Ub) is initially produced as a 229-a.a. precursor protein whose C-terminus Cys229 is post-translationally removed, followed by cleavages between a.a. 76 and 77, as well as between a.a. 152 and 153 to yield three idential 76-amino acid Ub molecules. Ub is involved in one of the most common post-translational modifications (PTMs) of cellular proteins, where Ub is linked covalently via its carboxyl terminus to usually Lys residues in target proteins. A given target Lys residue can be linked to one single Ub molecule (monoubiquitylated) or to a chain of Ub molecules (polyubiquitylated). In a polyubiquitin chain, ubiquitin molecules can be linked through one of the seven ubiquitin Lys residues (K6, K11, K27, K29, K33, K48, and K63) or through the Ub N-terminus Met1 residue (which generates linear chains). An Ub chain with a single linkage type is referred to as homotypic, whereas an heterotypic chain contains mixed linkages within the same chain, which can lead to branched/forked (as a result of one Ub linked to two or more Ubs) or non-branched Ub chains. Lys48-linked chains mark proteins for proteasomal degradation, while Lys63-linked chains regulate the activity/function of cellular proteins.
Specifications
Specifications
Ubiquitin, Lys48-Specific,
Apu2
Purified rabbit antibody conjugate in PBS with 15 mg/ml BSA and 0.1% sodium azide.
Purified
Rabbit
IgG
100μL
Signaling
Primary
NP_001268645.1
Human, Murine, Rat
Immunocytochemistry
Alexa Fluor 488
P0CG47
UBB
Immunogen used was human di-ubiquitin linked at Lys48.
Protein A purified
RUO
2°C to 8°C for one year from date of shipment
Monoclonal
Recognizes ubiquitin linked at Lys48. Does not recognize ubiquitin linked at Lys63 or free ubiquitin.