Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Ubiquitin, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP123015

Catalog No. NBP123015


Add to cart

Description

Description

UBA52 Polyclonal antibody specifically detects UBA52 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Ubiquitin
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
Affinity Purified
RPS27A
The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Ubiquitin using the numbering given in entry P62988.(GeneID 7311).
Immunogen affinity purified
RUO
Primary
6233
Immunohistochemistry (Paraffin)
Immunohistochemistry-Paraffin 1:100-1:500
P62988
RPS27A, UBA52, UBB ubiquitin B, UBC
Rabbit
IgG
0.1 ml
Store at 4C. Do not freeze.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only