Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant UCHL1.

Manufacturer:  Abnova Corporation H00007345A02

Catalog No. 89-004-359


Add to cart

Description

Description

The protein encoded by this gene belongs to the peptidase C12 family. This enzyme is a thiol protease that hydrolyzes a peptide bond at the C-terminal glycine of ubiquitin. This gene is specifically expressed in the neurons and in cells of the diffuse neuroendocrine system. Mutations in this gene may be associated with Parkinson disease

Sequence: NNQDKLGFEDGSVLKQFLSETEKMSPEDRAKCFEKNEAIQAAHDAVAQEGQCRVDDKVNFHFILFNNVDGHLYELDGRMPFPVNHGASSE
Specifications

Specifications

ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase)
Unconjugated
50% glycerol
NM_004181
PARK5|PGP9.5|Uch-L1
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
7345
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant UCHL1.
UCHL1
Serum
UCHL1
UCHL1 (NP_004172, 101 a.a. ~ 190 a.a) partial recombinant protein with GST tag.
RUO
Y
Polyclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit