Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCK2, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127835

Catalog No. PIPA127835

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Phosphorylates uridine and cytidine to uridine monophosphate and cytidine monophosphate. Does not phosphorylate deoxyribonucleosides or purine ribonucleosides. Can use ATP or GTP as a phosphate donor. Can also phosphorylate cytidine and uridine nucleoside

PA1-27835 detects UCK2 from human, mouse, and rat samples.PA1-27835 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Uridine-cytidine kinase 2.
Specifications

Specifications

UCK2
Polyclonal
PBS with no preservative
Q9BZX2, Q99PM9, Q9QYG8
TSA903, UK, UMPK
Chicken
IgY
50μg
Store at -20°C
Primary
7371
Western Blot
Unconjugated
UCK2
Purified
UCK2
Synthetic peptide corresponding to human Uridine-cytidine kinase 2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit