Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2V1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112841

Catalog No. 01-675-832


Add to cart

Description

Description

A suggested positive control for this product is Jurkat cell lysate.

Anti-Ubc13 is a monoclonal antibody specific to the ~19 kDa Ubc13 protein. Ubc13 is a ubiquitin conjugating enzyme that is complexed with a ubiquitin activating enzyme (E1), Mms2. The association of Mms2 to Ubc13 forms an active heterodimer that is involved in error-free post replicative DNA repair in yeast. Upon complexing with another conjugating enzyme variant, Uev1a, Ubc13 participates in the regulation of NF-kB signal transduction pathway in humans.
Specifications

Specifications

UBE2V1
Polyclonal
PBS with 0.02% sodium azide
Q13404, Q9CZY3
UBE2V1, ubiquitin-conjugating enzyme E2 variant 1, CIR1, UEV1, CROC1, UBE2V, UEV-1, UBE2V1, CROC-1, DNA-binding protein
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
296390, 66589, 7335
Western Blot
Unconjugated
UBE2V1
Liquid
UBE2V1
Synthetic peptide corresponding to 15 residues near the C-terminus of human UEV1A.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit