Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Ufc1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220801
Description
Ufc1 Polyclonal specifically detects Ufc1 in Human samples. It is validated for Western Blot.Specifications
Ufc1 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
HSPC155, ubiquitin-fold modifier conjugating enzyme 1, ubiquitin-fold modifier-conjugating enzyme 1, Ufm1-conjugating enzyme 1 | |
Rabbit | |
19 kDa | |
0.1 mL | |
Primary | |
Human. Species reactivity based on sequence identity: Mouse (96%), Cow (99%), Rat (98%), Zebrafish (89%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
0.8mg/mL | |
Western Blot 1:500-1:3000 | |
Q9Y3C8 | |
UFC1 | |
Recombinant protein encompassing a sequence within the center region of human UFC1. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
51506 | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only