Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ufd1L Mouse, Unlabeled, Clone: 19, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611642

Catalog No. BDB611642


Add to cart

Description

Description

Microdeletions of the chromosomal region 22q11.2 are associated with defects in cardiac and craniofacial development that are referred to as the CATCH 22 phenotype and include DiGeorge syndrome, the velo-cardio-facial syndrome, and the Opitz GBBB syndrome. Deleted in CATCH 22 patients, UFD1L encodes the human homolog of the yeast ubiquitin fusion degradation 1 protein (UFD1p), an essential component of the ubiquitin-dependent proteolytic pathway. The murine homolog (Ufd1l) is expressed in the eyes and in the inner ear primordia during embryogenesis. In rat liver cytosol, Ufd1l binds to the nuclear transport protein Npl4 and this binary complex competes with p47 for binding to p97, which inhibits Golgi membrane fusion. The tertiary complex Ufd1l/Npl4/p97 may also be involved in mitotic ubiquitin-dependent processes. Both Ufd1l and Npl4 are found in the nucleus, while p97 is found in the cytoplasm and nucleus. In humans, Ufd1L is primarily expressed in adult heart, placenta, skeletal muscle, and pancreas, and fetal liver and kidney. Thus, Ufd1L is thought to be
involved in cell functions that are critical for normal cell development in the cardiac and craniofacial regions

Host Species: Mouse
Clone: 19
Isotype: IgG1
Species Reactivity [for Features Main]: Mouse
Immunogen: Mouse Ufd1L aa. 120-241

Immunofluorescence, Western Blotting

Specifications

Specifications

Ufd1L
19
Unconjugated
Affinity Purified
Mouse Ufd1L aa. 120-241
50μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit