Learn More
UFM1 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems AF8237
Catalog No. AF8237
Add to cart
This item is not returnable. View return policy
Description
Description
UFM1 Polyclonal antibody AF8237 Specifically detects UFM1 in Human samples. It is validated for Immunocytochemistry, Western Blotting
Specifications
Specifications
UFM1
LYOPH
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
P61960
bA131P10.1, BM-002, C13orf20, chromosome 13 open reading frame 20, ubiquitin-fold modifier 1
Sheep
IgG
100 ug
Primary
51569
Reconstitute at 0.2 mg/mL in sterile PBS.
Immunocytochemistry, Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
UFM1
E. coli-derived recombinant human UFM1 Met1-Gly83 Accession # P61960
Immunogen affinity purified
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human UFM1 in direct ELISAs and Western blots.
Human