Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UGT3A2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523863

Catalog No. PIPA523863


Add to cart

Specifications

Specifications

UGT3A2
Polyclonal
Unconjugated
UGT3A2
Liquid
UGT3A2
KLH conjugated synthetic peptide between 377-405 amino acids from the C-terminal region of human UGT3A2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q3SY77, Q8JZZ0
UDP-glucuronosyltransferase 3A2, UDPGT 3A2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
167127, 223337
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit