Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ULK2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515381

Catalog No. PIPA515381

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

Macroautophagy is the major inducible pathway for the general turnover of cytoplasmic constituents in eukaryotic cells, it is also responsible for the degradation of active cytoplasmic enzymes and organelles during nutrient starvation. Macroautophagy involves the formation of double-membrane bound autophagosomes which enclose the cytoplasmic constituent targeted for degradation in a membrane bound structure, which then fuse with the lysosome (or vacuole) releasing a single-membrane bound autophagic bodies which are then degraded within the lysosome (or vacuole). Two human homologs of the yeast autophagy-specific kinase exist: ULK1(APG1) and ULK2. APG1 plays a critical role in regulating key elements of the autophagy pathway. APG1 stimulates autophagy, leading to autophagy-dependent restriction of cell growth and ultimately cell apoptosis at high levels of activity, and is a negative regulator of mTOR signaling.
Specifications

Specifications

ULK2
Polyclonal
PBS with 0.09% sodium azide
Q8IYT8
Serine/threonine-protein kinase ULK2; unc-51-like kinase 2; KIAA0623
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9706
Immunohistochemistry, Western Blot
Unconjugated
ULK2
Liquid
ULK2
KLH conjugated synthetic peptide between 235-264 amino acids from the N-terminal region of human ULK2
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit