Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UME6, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127859

Catalog No. PIPA127859

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Component of the RPD3C(L) histone deacetylase complex (HDAC) responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A. H2B. H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an i

PA1-27859 detects UME6 from yeast samples.PA1-27859 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27936 immunogen is fragment of yeast Transcriptional regulatory protein UME6.
Specifications

Specifications

UME6
Polyclonal
PBS with no preservative
P39001
UME6
Fragment of yeast Transcriptional regulatory protein UME6.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Yeast
Western Blot
Unconjugated
UME6
Purified
Chicken
IgY
50μg
Store at -20°C
Primary
851788
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit