Learn More
Unc84B/SUN2 Rabbit anti-Human, Canine, Horse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061038
Catalog No. 06-103-8MI
Add to cart
Description
Description
This Anti-Unc84B/SUN2 Antibody is validated for use in Immunoprecipitation and Western Blotting for the detection of Unc84B/SUN2.
Specifications
Specifications
Unc84B/SUN2
0.5mg/mL
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
FRIGG; UNC84B; RAB5IP; Rab5IP; SUN2
KLH-conjugated linear peptide corresponding to human Unc84B/SUN2 at and around the N-terminus.
Affinity Purfied
2-8°C for 12 months
Polyclonal
Canine, Equine, Human
Immunoprecipitation, Western Blot
Unconjugated
Affinity Purified
Rabbit
IgG
100μg
Primary
NP_056189