Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UNG Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NB6001031SS
Description
UNG Polyclonal specifically detects UNG in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
UNG | |
Polyclonal | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
DGUEC 3.2.2, HIGM4EC 3.2.2.27, UDGDKFZp781L1143, UNG15, UNG1uracil-DNA glycosylase 1, uracil-DNA glycosylase 2, UNG2, uracil-DNA glycosylase | |
Rabbit | |
Protein G purified | |
RUO | |
Primary | |
The synthetic peptide sequence used for immunogen is 100% conserved between UNG1 and UNG2. Thus the antibody recognizes both UNG1 and UNG2. As UNG1 and UNG2 are not resolved as two species in gel electrophoresis (Huag et al., 1998), they are indistinguishable by western blot. The observed molecular weight of UNG (UNG1 and UNG2) on western blots may vary: 26 kDa, 27.5 kDa, 31 kDa, and 38 kDa forms have been described (Bharati et al., 1998; Slupphaug et al., 1993). Please see Kruman et al. (Fig. 1D, 2004) for an example of western blot in rat. This specificity of this antibody has been validated by antisense UNG oligonucleotide (Kruman et al. 2004). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1.0 mg/ml | |
Western Blot 2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (Yang et al (2005)), Immunohistochemistry-Paraffin 10 ug/ml | |
NP_550433 | |
UNG | |
This antibody was generated by immunizing rabbit with a synthetic peptide sequence CRHFSKTNELLQKSGKKP corresponding to amino acids 281-298 of human UNG1 (NP 003353.1) and amino acids 290-307 of human UNG2 (NP 550433.1). The peptide sequence used for immunogen is highly conserved between mammalian and invertabrates. | |
0.025 mg | |
Base Excision Repair, DNA Repair, Stem Cell Markers | |
7374 | |
Human, Mouse, Rat, Rabbit | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction