Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cyclin O Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA520263
Description
This antibody will recognize both isoforms of Cyclin-O. A suggested positive control is human bladder tissue lysate. PA5-20263 can be used with blocking peptide PEP-0378.
Cyclin O Polyclonal antibody specifically detects Cyclin O in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Cyclin O | |
Polyclonal | |
Unconjugated | |
CCNO | |
C86987; CCNO; Ccnu; CILD29; cyclin domain containing; cyclin O; cyclin U; cyclin-like uracil-DNA glycosylase; cyclin-O; E130318K11; RGD1565217; UDG2; Ung2; uracil DNA glycosylase 2; uracil-DNA glycosylase 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10309, 218630, 499528 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P0C242, P22674 | |
CCNO | |
a 13 amino acid synthetic peptide from near the center of human Cyclin O | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction