Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNK Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524557

Catalog No. PIPA524557

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

UNK
Polyclonal
PBS with 0.09% sodium azide
Q9C0B0, Q8BL48
ZC3H5, ZC3HDC5, RING finger protein unkempt homolog, zinc finger CCCH domain-containing protein 5, zinc finger CCCH type domain containing 5, zinc finger CCCH-type containing 5, zinc finger CCCH-type domain containing 5
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
217331, 85451
Flow Cytometry, Western Blot
Unconjugated
UNK
Liquid
UNK
KLH conjugated synthetic peptide between 224-253 amino acids from the N-terminal region of human UNK
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit