Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UCKL1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514012

Catalog No. PIPA514012

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

Uridine kinase is a key enzyme in the pyrimidine salvage pathway of all mammalian cells. URKL1 (uridine kinase-like 1) is highly similar in structure and sequence to uridine kinase.
Specifications

Specifications

UCKL1
Polyclonal
PBS with 0.09% sodium azide
Q9NWZ5
Uridine/cytidine kinase-like 1; UCKL1; URKL1;
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54963
Immunohistochemistry, Western Blot
Unconjugated
UCKL1
Liquid
UCKL1
KLH conjugated synthetic peptide between 60-90 amino acids from the N-terminal region of human URKL1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit