Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

URM1, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA131921

Catalog No. PIPA131921

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-31921 detects Urm1 from yeast samples.PA1-31921 has been successfully used in Western blot applications.The PA1-31921 immunogen is: Recombinant full length protein (S. cerevisiae).
Specifications

Specifications

Urm1
Polyclonal
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
P40554
RP11-339B21.4, C9orf74, MGC2668
Rabbit
IgG
500μg
Store at 2°-8°C for up to 1 month. For long term storage store at -20°C
Primary
854809
ELISA, Western Blot
Unconjugated
URM1
Purified
URM1
Recombinant full length protein (S. cerevisiae).
IgG fraction
RUO
Antibody
Polyclonal
Yeast
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit