Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP22 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149644B
Description
USP22 Polyclonal specifically detects USP22 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
USP22 | |
Polyclonal | |
Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Q9UPT9 | |
USP22 | |
A genomic peptide made to an internal region of the human USP22 protein (within residues 100-205). [Swiss-Prot Q9H3R0] | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Signal Transduction, Stem Cell Markers | |
23326 | |
Store at 4C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Biotin | |
PBS with 0.05% Sodium Azide | |
Deubiquitinating enzyme 22, EC 3.1.2.15, EC 3.4.19.12, KIAA1063ubiquitin carboxyl-terminal hydrolase 22, KIAA1064, ubiquitin specific peptidase 22, ubiquitin specific peptidase 3-like, ubiquitin specific protease 22, ubiquitin thioesterase 22, Ubiquitin thiolesterase 22, ubiquitin-specific processing protease 22, Ubiquitin-specific-processing protease 22, USP3L | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction