Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ubiquitin specific peptidase 24 (B02), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human USP24 protein.

Manufacturer:  Abnova Corporation H00023358B02

Catalog No. 89-007-266


Add to cart

Description

Description

Modification of cellular proteins by ubiquitin is an essential regulatory mechanism controlled by the coordinated action of multiple ubiquitin-conjugating and deubiquitinating enzymes. USP24 belongs to a large family of cysteine proteases that function as deubiquitinating enzymes (Quesada et al., 2004 [PubMed 14715245]).[supplied by OMIM

Sequence: MISFLLGASRQNNQIRRWSSAQAREFGNLHNTVALLVLHSDVSSQRNVAPGIFKQRPPISIAPSSPLLPLHEEVEALLFMSEGKPYLLEVMFALRELTGSLLALIEMVVYCCFCNEHFSFTMLHFIKNQLETAPPHELKNTFQLLHEILVIEDPIQVERVKFVFETENGLLALMHHSNHVDSSRCYQCVKFLVTLAQKCPAAKEYFKENSHHWSWAVQWLQKKMSEHYWTPQSNVSNETSTGKTFQRTISAQDTLAYATALLNEKEQSGSSNGSESSPANENGDRHLQQGSESPMMIGELRSDLDDVDP
Specifications

Specifications

ubiquitin specific peptidase 24
Unconjugated
No additive
BC029660.1
FLJ31309|KIAA1057
Mouse
50 uL
Ubiquitin/Proteasome
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human USP24 protein.
USP24
Serum
USP24
USP24 (AAH29660.1, 1 a.a. ~ 309 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
23358
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit