Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP28 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA528072
Description
Recommended positive controls: A431. Predicted reactivity: Mouse (100%), Rat (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
USP28 Polyclonal antibody specifically detects USP28 in Human samples. It is validated for Western Blot.Specifications
USP28 | |
Polyclonal | |
Unconjugated | |
USP28 | |
9830148O20Rik; AU022237; Deubiquitinating enzyme 28; Kiaa1515; mKIAA1515; ubiquitin carboxyl-terminal hydrolase 28; ubiquitin carboxyl-terminal hydrolase 28 variant 1; ubiquitin specific peptidase 28; ubiquitin specific protease 28; ubiquitin thioesterase 28; ubiquitin thiolesterase 28; ubiquitin-specific-processing protease 28; Usp28 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57646 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q96RU2 | |
USP28 | |
Synthetic peptide corresponding to a region within amino acids 1015 and 1077 of Human USP28 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction