Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ubiquitin specific peptidase 3, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against a full-length human USP3 protein.

Manufacturer:  Abnova Corporation H00009960D01P

Catalog No. 89-018-190


  Check Availability
Add to cart

Specifications

Description

Sequence: MECPHLSSSVCIAPDSAKFPNGSPSSWCCSVCRSNKSPWVCLTCSSVHCGRYVNGHAKKHYEDAQVPLTNHKKSEKQDKVQHTVCMDCSSYSTYCYRCDDFVVNDTKLGLVQKVREHLQNLENSAFTADRHKKRKLLENSTLNSKLLKVNGSTTAICATGLRNLGNTCFMNAILQSLSNIEQFCCYFKELPAVELRNGKTAGRRTYHTRSQGDNNVSLVEEFRKTLCALWQGSQTAFSPESLFYVVWKIMPNFRGYQQQDAHEFMRYLLDHLHLELQGGFNGVSRSAILQENSTLSASNKCCINGASTVVTAIFGGILQNEVNCLICGTESRKFDPFLDLSLDIPSQFRSKRSKNQENGPVCSLRDCLRSFTDLEELDETELYMCHKCKKKQKSTKKFWIQKLPKVLCLHLKRFHWTAYLRNKVDTYVEFPLRGLDMKCYLLEPENSGPESCLYDLAAVVVHHGSGVGSGHYTAYATHEGRWFHFNDSTVTLTDEETVVKAKAYILFYVEHQAKAGSDKL
Description & Specifications

Specifications

ubiquitin specific peptidase 3
Unlabeled
In 1X PBS, pH 7.4
NM_006537.2
MGC129878, MGC129879, SIH003, UBP
Rabbit
Protein A column
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Western Blot
ubiquitin specific peptidase 3
USP3
Affinity Purified
USP3
USP3 (NP_006528.2, 1 a.a. ∽520 a.a) full-length human protein.
100μg
Ubiquitin/Proteasome
Primary
9960
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit