Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-USP35, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128733

Catalog No. NBP128733


Add to cart

Description

Description

USP35 Polyclonal antibody specifically detects USP35 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

USP35
1.0 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 10 ug/mg lysate
CAE51937.1
Deubiquitinating enzyme 35, EC 3.4.19.12, KIAA1372, ubiquitin carboxyl-terminal hydrolase 35, ubiquitin specific peptidase 35, ubiquitin specific protease 35, ubiquitin thioesterase 35, Ubiquitin thiolesterase 35, Ubiquitin-specific-processing protease 35, USP34
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, White-tufted-ear marmoset, Crab-eating macaque, African elephant, Naked mole rat, Small-eared galago and Thirteen-lined ground squirrel.
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
USP35
The immunogen recognized by this antibody maps to a region between residue 967 and 1017 of human ubiquitin specific peptidase 35 using the numbering given in entry CAE51937.1 (GeneID 57558).
Immunogen affinity purified
RUO
Primary
57558
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only