Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

USP4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527869

Catalog No. PIPA527869


Add to cart

Description

Description

PA5-27869 targets USP4 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-27869 immunogen is recombit fragment corresponding to a region within amino acids 528 and 872 of Human USP4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

USP4
Polyclonal
Unconjugated
USP4
Liquid
USP4
Recombit fragment corresponding to a region within amino acids 528 and 872 of Human USP4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q13107, P35123
UNP, Unph, deubiquitinating enzyme 4, ubiquitin carboxyl-terminal esterase 4, ubiquitin carboxyl-terminal hydrolase 4, ubiquitin specific protease 4 (proto-oncogene), ubiquitin thioesterase 4, ubiquitin thiolesterase 4, ubiquitin-specific processing protease 4, ubiquitous
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
22258, 7375
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit