Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP49 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525840
Description
Specifications
USP49 | |
Polyclonal | |
USP49 | |
deubiquitinating enzyme 49, ubiquitin carboxyl-terminal hydrolase 49, ubiquitin specific protease 49, ubiquitin thioesterase 49, ubiquitin thiolesterase 49, ubiquitin-specific-processing protease 49 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
224836, 25862 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q70CQ1, Q6P9L4 | |
USP49 | |
KLH conjugated synthetic peptide between 33-61 amino acids from the N-terminal region of human USP49 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction