Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ubiquitin specific peptidase 51, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant USP51.

Manufacturer:  Abnova Corporation H00158880A01

Catalog No. 89-004-806


Add to cart

Description

Description

Sequence: KHIHKHAETKQHHLAVDLYHGVIYCFMCKDYVYDKDIEQIAKETKEKILRLLTSTSTDVSHQQFMTSGFEDKQSTCE
Specifications

Specifications

ubiquitin specific peptidase 51
Unconjugated
50% glycerol
NM_201286
USP51
USP51 (NP_958443, 261 a.a. ~ 337 a.a) partial recombinant protein with GST tag.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
158880
ELISA, Western Blot
Mouse polyclonal antibody raised against a partial recombinant USP51.
USP51
Serum
Mouse
50 uL
Ubiquitin/Proteasome
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit