Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-USP9x, Alexa Fluor 700, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP148321AF700

Catalog No. NBP148321Z

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

USP9x
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
USP9X
Genomic peptide made to an internal region of the human USP9x protein (within residues 1150-1300). [Swiss-Prot Q93008]
270 kDa
0.1 ml
Apoptosis, Autophagy, Cancer, Neuroscience
Primary
8239
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Q93008
Deubiquitinating enzyme FAF-X, DFFRXubiquitin specific protease 9, X-linked (fat facets-like, Drosophila), Drosophila fat facets related, X-linked, EC 3.1.2.15, EC 3.4.19.12, FAF, FAM, Fat facets in mammals, fat facets protein related, X-linked, Fat facets protein-related, X-linked, hFAM, probable ubiquitin carboxyl-terminal hydrolase FAF-X, ubiquitin specific peptidase 9, X-linked, ubiquitin specific peptidase 9, X-linked (fat facets-like, Drosophila), ubiquitin thioesterase FAF-X, Ubiquitin thiolesterase FAF-X, ubiquitin-specific processing protease FAF-X, Ubiquitin-specific protease 9, X chromosome, Ubiquitin-specific-processing protease FAF-X, USP9, X chromosome (fat facets-like Drosophila)
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human and mouse. Immunogen sequence has 95% homology to rat and 80% homology to Zebrafish.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only