Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UTP6 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521716

Catalog No. PIPA521716


Add to cart

Description

Description

PA5-21716 targets UTP6 in ICC, IF, and WB applications and shows reactivity with Human samples. The PA5-21716 immunogen is a recombit fragment corresponding to a region within amino acids 213 and 597 of UTP6 (Uniprot IDNo.Q9NYH9). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

UTP6
Polyclonal
Unconjugated
UTP6
Liquid
UTP6
Recombit fragment corresponding to a region within amino acids 213 and 597 of UTP6 (Uniprot IDNo.Q9NYH9)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9NYH9
C17orf40, HCA66, U3 small nucleolar RNA-associated protein 6 homolog, hepatocellular carcinoma associated antigen 66, hepatocellular carcinoma-associated antigen 66, multiple hat domains protein
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
55813
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit